Learn More
Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas
Click to view available options
Length (Metric):
100 mm
130 mm
150 mm
185 mm
210 mm
Length (Metric) Blade:
30 mm
35 mm
40 mm
50 mm
55 mm
Width (Metric) Blade:
2 mm
3 mm
4 mm
5 mm
6 mm
9 mm
Unit Size:
Each
Description
- Double endet
- Stainless steel
- Autoclavable
- Straight
Specifications
Specifications
| Quantity | 1 Ea. |
| Autoclavable | Autoclavable |
| Shape | Straight |
| Product Type | Double Ended Micro Spatula |
| For Use With (Application) | Multi-purpose spatula for a wide range of uses |
| Length (Metric) Blade | 55 mm |
| Width (Metric) | 6 mm |
| Width (Metric) Blade | 6 mm |
| Blade Material | Stainless Steel (18/10) |
| Diameter (Metric) Outer | 2mm |
| Show More |