Learn More
Agencinox™ 18/10 Stainless Steel Evaporating Dish
Click to view available options
Capacity (Metric):
50 mL
100 mL
Unit Size:
Each
Description
- Low form evaporating dish
- Stainless steel construction
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 50 mL |
| Material | Stainless Steel |
| Shape | Round |
| Type | Evaporating Dish |
| For Use With (Application) | Evaporating Application |
| Height (Metric) | 20 mm |
| Diameter (Metric) | 60 mm |