Learn More
Bochem™ 18/10 Stainless Steel Evaporating Dish
Product Code. p-8006739
Click to view available options
Height (Metric):
20 mm
35 mm
40 mm
Unit Size:
Each
Product Code. 10160411
Brand: Bochem™ 8454
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Bottom Shape Flat
Material Stainless Steel
Shape Rectangular
Type Evaporating Dish
Length (Metric) 220 mm
Height (Metric) 40 mm
Weight (Metric) 260 g