Learn More
Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim
Product Code. p-8003315
Click to view available options
Height (Metric):
45 mm
60 mm
95 mm
100 mm
Length (Metric):
310 mm
340 mm
375 mm
445 mm
500 mm
Unit Size:
Each
Product Code. 10346104
Brand: Bochem™ 8399
Only null left
Add to basket
Add to basket
Evaporate materials in the laboratory wit the Bochem™ 18/10 Stainless Steel Evaporating Dish with Rim.
  • 18/10 stainless steel
  • Substantial rim size
  • Rectangular shape
Specifications
Bottom Shape Flat
Material Stainless Steel
Shape Rectangular
Type Evaporating Dish
Length (Metric) 500 mm
Height (Metric) 100 mm