Learn More
Bochem™ Stainless Steel Ladle Scoop
Product Code. 10434881
Click to view available options
Length (Metric):
350 mm
Unit Size:
Each
Product Code. 10434881
Brand: Bochem™ 8744
Only null left
Add to basket
Add to basket
Flat-handled and constructed of 18/10 stainless steel. Bochem™ Stainless Steel Ladle Scoop is durable and resistant.
  • 18/10 stainless steel construction.
  • Flat-handle design.
Specifications
Quantity 1 Ea.
Capacity (Metric) 1000 mL
Product Type Ladle Scoop
Length (Metric) Handle 350 mm
Handle Material Stainless Steel (18/10)
Material Stainless Steel (18/10)
Handle Type Flat
Length (Metric) 350 mm