Learn More
RSG Rostfrei Schneidwerkzeuge™ Stainless Steel Micro Spatulas
Product Code. 11876890
Encompass
Click to view available options
Length (Metric):
150 mm
Unit Size:
Pack of 5
Product Code. 11876890
Brand: RSG Rostfrei Schneidwerkzeuge™ 87374
This item requires a price update prior to being ordered.
Request a Price for this item here.
Excellent for scraping, transferring or applying chemicals or treatments in the laboratory. RSG Rostfrei Schneidwerkzeuge Stainless Steel Spatulas are durable and chemically resistant.
  • With plastic handles
  • Available in a variety of end styles
Specifications
Quantity 5 Pk.
Shape Single
Product Type Micro Spatula
Handle Color Black
Width (Metric) 4 mm
Handle Material Plastic
Material Stainless Steel (18/10)
Handle Type Plastic
Length (Metric) 150 mm