missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Spatulas, Spoon/round/bent

Manufactured from 18/10 stainless steel

Brand:  Bochem™ 3202

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10663541

  • 2.20€ / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

  • Made from 18/10 stainless steel
Specifications

Specifications

1 Ea.
Spoon/Round/Bent
Used for a variety of applications
2mm
210 mm
Autoclavable
Spatula
Stainless Steel (18/10)
Stainless Steel (18/10)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers