Learn More
Bochem™ 18/10 Stainless Steel Spoon Spatulas
Product Code. 10549471
Click to view available options
Length (Metric):
150 mm
180 mm
210 mm
250 mm
300 mm
Length (Metric) Blade:
40 mm
50 mm
70 mm
Unit Size:
Each
Pack of 10
Product Code. 10549471
Brand: Bochem™ 3473
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Spoon Spatulas serve a wide variety of functions in the lab.
  • Manufactured from 18/10 stainless steel
Specifications
Quantity 10 Pk.
Product Type Spoon Spatula
Length (Metric) Blade 50 mm
Width (Metric) 12 mm
Width (Metric) Blade 10 mm
Material Stainless Steel (18/10)
Length (Metric) 210 mm