missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetic Acid, TRC

Product Code. 30329535
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30329535

Brand: Toronto Research Chemicals TRCB6790751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromoacetic acid
CAS 79-08-3
Purity Grade Notes GC
Molecular Formula C2 H3 Br O2
Quantity 1 g
Synonym Acetic acid, bromo- (8CI,9CI), 2-Bromoacetic acid, 2-Bromoethanoic acid, Bromoacetate ion, Bromoacetic acid, Bromoethanoic acid, Carboxymethyl bromide, MBAA, Monobromoacetic acid, NSC 141, To NTu, α-Bromoacetic acid
InChI Formula InChI=1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)
SMILES OC(=O)CBr
IUPAC Name 2-bromoacetic acid
Molecular Weight (g/mol) 138.95
Formula Weight 137.9316
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.