missing translation for 'onlineSavingsMsg'
Learn More

Beclomethasone Dipropionate, TRC

Product Code. 30341598
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30341598

Brand: Toronto Research Chemicals TRCB1310301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beclometasone Dipropionate
CAS 5534-09-8
Purity Grade Notes HPLC
Molecular Formula C28 H37 Cl O7
Quantity 1 g
Synonym 9-Chloro-11beta-hydroxy-16beta-methyl-3,20-dioxopregna-1,4-diene-17,21-diyl Dipropanoate, Beclometasone Dipropionate, Betamethasone Dipropionate Imp. E (Pharmeuropa), (11β,16β)-9-Chloro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate, 9α-Chloro-16β-methylprednisolone 17,21-dipropionate, Aerobec, Aldecin, Aldecin AQ nasal, Anceron, Andion, Beclacin, Beclate, Beclazone, Beclazone 250, Beclazone 50, Beclomet, Beclometasone 17,21-dipropionate, Beclometasone dipropionate, Beclomethasone 17,21-dipropionate, Beclomethasone 17α,21-dipropionate, Beclomethasone dipropionate, Beclotide, Beclotide 100, Becloval, Beclovent, Beclovent Inhaler, Becodisks, Beconase, Beconase AQ, Becotide, Belchlorhinol, Belcoforte, Belcomet, Clenil A, Entyderma, Inalone O, Inalone R, Korbutone, Propaderm, Propaderm Forte, QVAR, Qvar 50, Rino-Clenil, Sanasthmax, Sanasthmyl, Sanasthymyl, Sch 8020W, Vancenase, Vancenase AQ, Vanceril, Vanceril DS, Ventolair, Viarex, Viarox, orBec
InChI Formula InChI=1S/C28H37ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1
SMILES CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C
IUPAC Name [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Molecular Weight (g/mol) 521.04
Formula Weight 520.2228
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.