missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Palmitate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Palmitic acid-n-butyl ester |
| CAS | 111-06-8 |
| Molecular Formula | C20 H40 O2 |
| Quantity | 1 g |
| Synonym | Palmitic acid-n-butyl ester, Hexadecanoic acid, butyl ester (9CI, ACI), Palmitic acid, butyl ester (6CI, 7CI, 8CI), Butyl hexadecanoate, Butyl palmitate, n-Butyl hexadecanoate, n-Butyl palmitate, NSC 4815, Palmitic acid n-butyl ester |
| InChI Formula | InChI=1S/C20H40O2/c1-3-5-7-8-9-10-11-12-13-14-15-16-17-18-20(21)22-19-6-4-2/h3-19H2,1-2H3 |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCCCC |
| IUPAC Name | butyl hexadecanoate |
| Molecular Weight (g/mol) | 312.53 |
| Formula Weight | 312.3028 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction