missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)aniline, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N-(tert-Butoxycarbonyl)aniline |
| CAS | 3422-01-3 |
| Molecular Formula | C11H15NO2 |
| Quantity | 1 g |
| Synonym | N-Phenyl-carbamic Acid 1,1-Dimethylethyl Ester, N-BOC-aniline, tert-Butyl N-Phenylurethane, tert-Butyl N-Phenylcarbamate, tert-Butyl Phenylcarbamate |
| InChI Formula | InChI=1S/C11H15NO2/c1-11(2,3)14-10(13)12-9-7-5-4-6-8-9/h4-8H,1-3H3,(H,12,13) |
| SMILES | CC(C)(C)OC(=O)Nc1ccccc1 |
| IUPAC Name | tert-butyl N-phenylcarbamate |
| Molecular Weight (g/mol) | 193.24 |
| Formula Weight | 193.1103 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction