Learn More
1H-Benzotriazole-(ring-d4) solution, Honeywell Fluka™
Product Code. 15900192 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15900192
Brand: Honeywell Fluka R13501ML
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10 μg/mL in acetone (15949681), 50 μg/mL in acetone (15900192) Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
CAS 67-64-1, 1185072-03-0
Density 0.79 g/cm3
Flash Point -17°C
Physical Form Liquid
Boiling Point 55°C to 57°C
Industry Type Parabenes
Quantity 1 mL
Grade Analytical Standard