Learn More
1H-Indene-3-carboxylic acid, 97%
Product Code. 15477277
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15477277 1 g 1g
15487277 5 g 5g
2 options
Product Code. 15477277
Brand: Thermo Scientific Alfa Aesar H34344.03
Request Bulk or Custom Format
CAS: 5020-21-3 | C10H8O2 | 160.172 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5020-21-3
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.172
MDL Number MFCD00086207
InChI Key NZSCUDBGUBVDLO-UHFFFAOYSA-N
Synonym 1h-indene-3-carboxylic acid, indene-3-carboxylic acid, 3-indenecarboxylic acid
PubChem CID 84261
IUPAC Name 3H-indene-1-carboxylic acid
SMILES C1C=C(C2=CC=CC=C21)C(=O)O
Specifications
Melting Point 161°C to 165°C
Quantity 1 g
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material 1H-Indene-3-carboxylic acid