Learn More
Chem-Lab 1LT Barium perchlorate 0.005 mol/l in iso-
Brand:  Chem-Lab CL05.0204.1000
1LT Barium perchlorate 0.005 mol/l in iso-Ppropanol (1.6812 g Ba(ClO4)2 / l C3H8O/H2O =,
Product Code. 11963103
  • 111.00€ / 1 L
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Barium perchlorate 0.005 mol/l in Propanol-2
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Barium perchlorate,Propanol-2