Learn More
Chem-Lab 1LT Barium perchlorate 0.005 mol/l in iso-
Brand: Chem-Lab CL05.0204.1000
1LT Barium perchlorate 0.005 mol/l in iso-Ppropanol (1.6812 g Ba(ClO4)2 / l C3H8O/H2O =,
Safety and Handling
- Barium perchlorate 0.005 mol/l in Propanol-2
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contain: Barium perchlorate,Propanol-2