Learn More
Chem-Lab 1LT Perchloric acid 60% ar (60+% HClO4)
Brand: Chem-Lab CL00.1615.1000
1LT Perchloric acid 60% ar (60+% HClO4)
Safety and Handling
- Perchloric acid 60% a.r.
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contain: Perchloric acid