Learn More
Chem-Lab 1LT Potassium bromate 1/60 mol/l (2.7835 g KBrO3/ l H2O = 0.1 N (±0.0002/20°C))
Brand: Chem-Lab CL05.1101.1000
1LT Potassium bromate 1/60 mol/l (2.7835 g KBrO3/ l H2O = 0.1 N (±0.0002/20°C))
Safety and Handling
- Potassium bromate 0.0167 mol/l
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Oxidising liquid Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Potassium bromate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.