missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagecon 1LT SODIUM LAURYL (DODECYL) S ULPHATE (0.1N) 1L
Brand: Reagecon SLS011
1LT SODIUM LAURYL (DODECYL) S ULPHATE (0.1N) 1L
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- SODIUM LAURYL (DODECYL) SULPHATE 0.1N (0.1M)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium dodecyl sulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction