missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetoxyvalerenic Acid, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Acetoxyvalerenic Acid |
| CAS | 84638-55-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H24 O4 |
| Quantity | 1 mg |
| Synonym | [1R-[1alpha,4alpha(E),7beta,7aalpha]]-3-[1-(Acetyloxy)-2,4,5,6,7,7a-hexahydro-3,7-dimethyl-1H-inden-4-yl]-2-methyl-2-propenoic acid, Acetylvalerenolic acid, Acetoxyvalerenic acid |
| InChI Formula | InChI=1S/C17H24O4/c1-9-5-6-13(7-11(3)17(19)20)15-10(2)8-14(16(9)15)21-12(4)18/h7,9,13-14,16H,5-6,8H2,1-4H3,(H,19,20)/b11-7+/t9-,13+,14-,16+/m1/s1 |
| SMILES | C[C@@H]1CC[C@@H](\C=C(/C)\C(=O)O)C2=C(C)C[C@@H](OC(=O)C)[C@H]12 |
| IUPAC Name | (E)-3-[(1R,4S,7R,7aR)-1-acetyloxy-3,7-dimethyl-2,4,5,6,7,7a-hexahydro-1H-inden-4-yl]-2-methylprop-2-enoic acid |
| Molecular Weight (g/mol) | 292.37 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction