missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amoxicillin-d4 (Major), TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Amoxicillin-d4 |
| CAS | 2673270-36-3 |
| Molecular Formula | C16 D4 H15 N3 O5 S |
| Quantity | 1 mg |
| Synonym | (2S,5R,6R)-6-[[(2R)-2-Amino-2-(4-hydroxy-2,3,5,6-tetradeuteriophenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, (2S,5R,6R)-6-[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid-d4, 6-[D-α-Amino-α-(4-hydroxyphenyl-d4)acetamido]penicillanic acid, Amoxicillin (phenyl-D4), Amoxicillin-D4 (4-hydroxyphenyl-D4), Amoxicillin D4 (4-hydroxyphenyl D4), AMPC-d4, Amoxycillin-d4, Helvamox-d4, Pasetocin-d4, Penimox-d4, Zamocillin-d4 |
| InChI Formula | InChI=1S/C16H19N3O5S/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24)/t9-,10-,11+,14-/m1/s1/i3D,4D,5D,6D |
| SMILES | [2H]c1c([2H])c([C@@H](N)C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)O)c([2H])c([2H])c1O |
| IUPAC Name | (2S,5R,6R)-6-[[(2R)-2-amino-2-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| Molecular Weight (g/mol) | 369.4288 |
| Formula Weight | 369.1296 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction