missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl)adipate-d8, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Bis(2-ethylhexyl)adipate-d8 |
| CAS | 1214718-98-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 D8 H34 O4 |
| Quantity | 1 mg |
| Synonym | Adipic Acid, Bis-2-ethylhexyl Ester D8 (adipate D8), Hexanedioic-2,2,3,3,4,4,5,5-d8 acid, 1,6-bis(2-ethylhexyl) ester (ACI), 1,6-Bis(2-ethylhexyl) hexanedioate-2,2,3,3,4,4,5,5-d8 (ACI) |
| InChI Formula | InChI=1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3/i11D2,12D2,15D2,16D2 |
| SMILES | [2H]C([2H])(C(=O)OCC(CC)CCCC)C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)OCC(CC)CCCC |
| IUPAC Name | bis(2-ethylhexyl) 2,2,3,3,4,4,5,5-octadeuteriohexanedioate |
| Molecular Weight (g/mol) | 378.616 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction