missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisoprolol-D5, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Bisoprolol-d5 |
| CAS | 1189881-87-5 |
| Molecular Formula | C18H26D5NO4 |
| Quantity | 1 mg |
| Synonym | 1-[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol-d5 |
| InChI Formula | InChI=1S/C18H31NO4/c1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4/h5-8,14-15,17,19-20H,9-13H2,1-4H3/i11D2,13D2,17D |
| SMILES | CC(C)OCCOCC1=CC=C(OC([2H])([2H])C(O)([2H])C([2H])([2H])NC(C)C)C=C1 |
| IUPAC Name | 1,1,2,3,3-pentadeuterio-1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol |
| Molecular Weight (g/mol) | 330.47 |
| Formula Weight | 330.2567 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction