missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CHIPOpt
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
CHIPOpt is a CHIP/STUB1 E3 ligase inhibitor. In vitro, CHIPOpt decreases phosphorylated tau ubiquitination, with little effect on unmodified tau. In Thioflavin T assays, CHIPOpt inhibits the ability of CHIP to suppress phosphorylated tau aggregation. FITC-CHIPOp (Cat. No. 8045), a fluorescent CHIPOpt, is also available.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | 757.84 |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction