Learn More
Enzo Life Sciences 1mg Okadaic acid (high purity)
Brand:  Enzo Life Sciences ALX-350-003-M001
Encompass_Preferred
1mg Okadaic acid (high purity)
Product Code. 15139007
  • 2545.84€ / 1mg
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Okadaic acid (high purity)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.