missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buflomedil Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Buflomedil Hydrochloride 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Buflomedil hydrochloride |
| CAS | 35543-24-9 |
| Analyte or Component Names | Buflomedil hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H25 N O4 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Buflomedil Hydrochloride, 4-(1-Pyrrolidinyl)-1-(2,4,6-trimethoxyphenyl)-1-butanone hydrochloride, Bufedil, Buflan, Buflocit, Buflomedil hydrochloride, Buflonat, Fonzylane, Irrodan, Irrodan R, LL 1656, Lofton, Loftyl, Provas |
| InChI Formula | InChI=1 S/C17H25NO4.ClH/c1-20-13-11-15(21-2)17(16(12-13)22-3)14(19)7-6-10-18-8-4-5-9-18;/h11-12 H,4-10H2,1-3H3;1 H |
| SMILES | Cl.COc1cc(OC)c(C(=O)CCCN2CCCC2)c(OC)c1 |
| IUPAC Name | 4-pyrrolidin-1-yl-1-(2,4,6-trimethoxyphenyl)butan-1-one;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction