missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Desmethylvenlafaxine, 1mg/ml in Methanol, Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
O-Desmethylvenlafaxine, 1mg/ml in Methanol
Specifications
Specifications
| Chemical Name or Material | O-Desmethylvenlafaxine |
| CAS | 93413-62-8 |
| Analyte or Component Names | O-Desmethylvenlafaxine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H25 N O2 |
| Concentration | 1 mg/mL |
| Synonym | O-Desmethylvenlafaxine, 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, D-VENIZ, DVS 233, Desvenlafaxine, Pristiq |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CN(C)CC(c1ccc(O)cc1)C2(O)CCCCC2 |
| IUPAC Name | 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction