missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sildenafil citrate, 1mg/ml in Methanol (as free base), Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Sildenafil citrate, 1mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Sildenafil Citrate |
| CAS | 171599-83-0 |
| Analyte or Component Names | Sildenafil Citrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H30 N6 O4 S . C6 H8 O7 |
| Concentration | 1 mg/mL |
| Synonym | Sildenafil Citrate, 7 H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Piperazine, 1-[[3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1 H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1) (9 CI), 1-[[3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1 H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Actra-Rx, Alisiga, Apodefil, Caverta 100, Edegra, Kamagra, Maxifort, Niagra Actra-Rx, Penegra, Sildegra, Sildenafil citrate, Sildigra, Siligra, Sollevare, Tonafil, UK 92480, UK 92480-10, Vega, Vega (pharmaceutical), Viagra, Vigrex, Zenegra |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(C)CC4.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| IUPAC Name | 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-6 H-pyrazolo[4,3-d]pyrimidin-7-one;2-hydroxypropane-1,2,3-tricarboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction