missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Chromium Copper, CDA 182, C18200, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Chromium Copper, CDA 182, C18200, Ø38x3mm
Specifications
Specifications
| CAS | 7440-50-8 |
| Form | Disc |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction