missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ni/Cr/Mo, AISI 8620, G86200, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Alloy Steel, Ni/Cr/Mo, AISI 8620, G86200, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Lead/Boron/Niobium/Zirconium/Antimony/Tungsten/Vanadium/Arsenic/Cobalt/Nitrogen/Tin/Phosphorus/Sulfur/Aluminium/Molybdenum/Copper/Carbon/Silicon/Chromium/Nickel/Manganese/Iron |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium -12 ppm/Lead -10 ppm/Boron -3 ppm/Niobium 0.0014 %/Zirconium 0.0014 %/Antimony 0.0024 %/Tungsten 0.003 %/Vanadium 0.0039 %/Arsenic 0.0045 %/Cobalt 0.0072 %/Nitrogen 0.0082 %/Tin 0.0094 %/Phosphorus 0.0128 %/Sulfur 0.0224 %/Aluminium 0.0241 %/Molybdenum 0.19 %/Copper 0.199 %/Carbon 0.211 %/Silicon 0.237 %/Chromium 0.507 %/Nickel 0.551 %/Manganese 0.811 %/Iron (97) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction