missing translation for 'onlineSavingsMsg'
Learn More

2,2,2,3'-Tetrafluoroacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7055977
23.26 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
708-64-5
C8H4F4O
192.11
MFCD00039236
NQSPTMFCJGKOQJ-UHFFFAOYSA-N
2,2,2,3'-tetrafluoroacetophenone, 2,2,2-trifluoro-1-3-fluorophenyl ethanone, m-fluorotrifluoroacetophenone, acetophenone, 2,2,2,3'-tetrafluoro, 2,2,2-trifluoro-1-3-fluorophenyl ethan-1-one, ethanone, 2,2,2-trifluoro-1-3-fluorophenyl, ethanone,2,2,2-trifluoro-1-3-fluorophenyl, acmc-20aol4, m,alpha,alpha,alpha-tetrafluoroacetophenone, 2,2,2,3'-trtrafluoroacetophenone
69723
2,2,2-trifluoro-1-(3-fluorophenyl)ethanone
FC1=CC=CC(=C1)C(=O)C(F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
708-64-5
C8H4F4O
192.11
MFCD00039236
NQSPTMFCJGKOQJ-UHFFFAOYSA-N
2,2,2,3'-tetrafluoroacetophenone, 2,2,2-trifluoro-1-3-fluorophenyl ethanone, m-fluorotrifluoroacetophenone, acetophenone, 2,2,2,3'-tetrafluoro, 2,2,2-trifluoro-1-3-fluorophenyl ethan-1-one, ethanone, 2,2,2-trifluoro-1-3-fluorophenyl, ethanone,2,2,2-trifluoro-1-3-fluorophenyl, acmc-20aol4, m,alpha,alpha,alpha-tetrafluoroacetophenone, 2,2,2,3'-trtrafluoroacetophenone
69723
2,2,2-trifluoro-1-(3-fluorophenyl)ethanone
FC1=CC=CC(=C1)C(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.372 g/mL
Boiling Point 79°C (50 mmHg)
Flash Point 17°C (63°F)
Refractive Index 1.444
Quantity 1 g
UN Number UN1224
Beilstein 1872788
Formula Weight 192.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,2,3'-Tetrafluoroacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'