missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,2,2-Trifluoroethyl trifluoromethanesulfonate, 95%

Brand:  Alfa Aesar™ H61603.14

Additional Details : CAS : 6226-25-1 Weight : 0.02500kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 15406646

  • 383.00€ / 25g


In stock, ready to ship: 1
Add to basket

Description

Description

Applications
2,2,2-Trifluoroethyl trifluoromethanesulfonate is a powerful trifluoroethylating agent which is useful for synthesis of fluorinated amino acids. It is used as a reagent in thenantioselective preparation of cyclic N-aryl hydroxamic acids via phase-transfer catalyzed alkylation of nitrobenzyl bromides to give nirophenylalanines.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

6226-25-1
232.096
RTMMSCJWQYWMNK-UHFFFAOYSA-N
80366
C(C(F)(F)F)OS(=O)(=O)C(F)(F)F
C3H2F6O3S
MFCD00671579
2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
2,2,2-trifluoroethyl trifluoromethanesulfonate

Specifications

2,2,2-Trifluoroethyl trifluoromethanesulfonate
>110°C (230°F)
Moisture sensitive
25g
C3H2F6O3S
MFCD00671579
2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
RTMMSCJWQYWMNK-UHFFFAOYSA-N
2,2,2-trifluoroethyl trifluoromethanesulfonate
80366
Liquid
1.611 g/mL
1.306
Colorless to Yellow
6226-25-1
CF3SO3CH2CF3
UN3265
Slightly soluble in water.
C(C(F)(F)F)OS(=O)(=O)C(F)(F)F
232.096
232.1
95%
Videos
SDS Documents