missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2′-Bipyridyl, ≥99.0% (NT), Honeywell Fluka™
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 69°C to 71°C |
Boiling Point | 273°C |
UN Number | UN2811 |
Beilstein | 113089 |
Formula Weight | 156.18g/mol |
Percent Purity | ≥99.0% (NT) |
Packaging | Plastic Bottle |
Quantity | 25 g |
Chemical Name or Material | 2,2′-Bipyridyl |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'