missing translation for 'onlineSavingsMsg'
Learn More

2-(2-chlorophenoxy)ethylamine, 97%, Thermo Scientific™

Product Code. p-2696061
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10181032

Brand: Thermo Scientific Maybridge RF03300CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 26378-53-0
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.624
MDL Number MFCD00125294
InChI Key NAPNYPMMDVRKGM-UHFFFAOYSA-N
PubChem CID 2735788
IUPAC Name 2-(2-chlorophenoxy)ethanamine
SMILES C1=CC=C(C(=C1)OCCN)Cl

Specifications

CAS Min % 97.0
Quantity 250 mg
Packaging Amber Glass Bottle
Assay Percent Range 97%
Linear Formula ClC6H4O(CH2)2NH2
Synonym 2-2-chlorophenoxy ethylamine, 2-2-chlorophenoxy ethanamine, 2-2-chlorophenoxy-1-ethanamine, ethanamine, 2-2-chlorophenoxy, 2-2-chlorophenoxy ethyl amine, 1-2-aminoethoxy-2-chlorobenzene, acmc-20aoqz, ethanamine,2-2-chlorophenoxy, 2-2-chlorophenoxy ethan-1-amine
Formula Weight 171.63
Percent Purity 97%
Chemical Name or Material 2-(2-Chlorophenoxy)ethylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.