missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals

Catalog Number p-7031100
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
2032-75-9
C7H4Cl2O2
191.01
MFCD00196818
NMNKOUVBELKKTR-UHFFFAOYSA-N
1,3-benzodioxole,2,2-dichloro, 2,2-dichlorobenzo d 1,3 dioxole, dcmdb, 1,2-dichloromethylenedioxy benzene
11041568
2,2-dichloro-2H-1,3-benzodioxole
ClC1(Cl)OC2=CC=CC=C2O1
This item is not returnable. View return policy
2032-75-9
C7H4Cl2O2
191.01
MFCD00196818
NMNKOUVBELKKTR-UHFFFAOYSA-N
1,3-benzodioxole,2,2-dichloro, 2,2-dichlorobenzo d 1,3 dioxole, dcmdb, 1,2-dichloromethylenedioxy benzene
11041568
2,2-dichloro-2H-1,3-benzodioxole
ClC1(Cl)OC2=CC=CC=C2O1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.54
Boiling Point 215°C
Flash Point 85°C (185°F)
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 191.01
Percent Purity 97%
Chemical Name or Material 2,2-Dichloro-1,3-benzodioxole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'