missing translation for 'onlineSavingsMsg'
Learn More

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7031836
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
128073-33-6
C5H6F2O2
136.098
MFCD07777163
HLFLYOQLHYYNLT-UHFFFAOYSA-N
2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid
14695229
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
CC1(CC1(F)F)C(=O)O
This item is not returnable. View return policy
128073-33-6
C5H6F2O2
136.098
MFCD07777163
HLFLYOQLHYYNLT-UHFFFAOYSA-N
2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacid
14695229
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
CC1(CC1(F)F)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 62°C to 64°C
Quantity 250 mg
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material (±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'