missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorocyclopropylamine hydrochloride, 97%

Product Code. 11452220
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
11452220 500 mg 500mg
11442220 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 11452220

Brand: Thermo Scientific Alfa Aesar H29051.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 105614-25-3 | C3H6ClF2N | 129.535 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105614-25-3
Molecular Formula C3H6ClF2N
Molecular Weight (g/mol) 129.535
MDL Number MFCD07777150
InChI Key WSSNWLHFBOKGGL-UHFFFAOYSA-N
Synonym 2,2-difluorocyclopropylamine hydrochloride, 2,2-difluorocyclopropan-1-amine hydrochloride, 2,2-difluorocycloprop-1-ylamine hydrochloride, 2,2-difluorocyclopropanamine hydrochloride, cyclopropanamine, 2,2-difluoro-, hydrochloride, 2,2-difluorocyclopropanamine, acmc-20aojv, 2,2-difluorocyclopropylamine, chloride, 2,2-difluorocycloprop-1-ylaminehydrochloride, 2,2-difluoro-1-cyclopropanamine hydrochloride
PubChem CID 20161313
IUPAC Name 2,2-difluorocyclopropan-1-amine;hydrochloride
SMILES C1C(C1(F)F)N.Cl

Specifications

Melting Point ∽145°C (decomposition)
Quantity 500 mg
Sensitivity Hygroscopic
Formula Weight 129.54
Percent Purity 97%
Chemical Name or Material 2,2-Difluorocyclopropylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.