missing translation for 'onlineSavingsMsg'
Learn More

(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Dibenzyl Carbamate Phosphate, TRC

Product Code. 30383418
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383418 50 mg 50mg
30371126 100 mg 100mg
30388014 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383418 Supplier Toronto Research Chemicals Supplier No. TRCD48484050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Dibenzyl Carbamate Phosphate
Analyte or Component Names (2,2-Dimethyl-1,3-dioxolan-4-yl)methyl Dibenzyl Carbamate Phosphate
Shipping Condition Room Temperature
Molecular Formula C26 H35 N2 O10 P
InChI Formula InChI=1S/C26H35N2O10P/c1-26(2)34-19-23(38-26)20-37-39(31,35-15-13-27-24(29)32-17-21-9-5-3-6-10-21)36-16-14-28-25(30)33-18-22-11-7-4-8-12-22/h3-12,23H,13-20H2,1-2H3,(H,27,29)(H,28,30)
Solution Type Neat
SMILES CC1(C)OCC(COP(=O)(OCCNC(=O)OCc2ccccc2)OCCNC(=O)OCc3ccccc3)O1
IUPAC Name benzyl N-[2-[(2,2-dimethyl-1,3-dioxolan-4-yl)methoxy-[2-(phenylmethoxycarbonylamino)ethoxy]phosphoryl]oxyethyl]carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 566.537
Quantity 50 mg
Formula Weight 566.203
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.