missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-2,3-dihydro-1-benzofuran-5-thiol, TRC

Product Code. 30373624
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30373624 50 mg -
30379755 250 mg -
30377822 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 30373624

Brand: Toronto Research Chemicals TRCD46154850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1248920-72-0
Molecular Formula C10H12OS
Molecular Weight (g/mol) 180.27
SMILES CC1(C)Cc2cc(S)ccc2O1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H12OS/c1-10(2)6-7-5-8(12)3-4-9(7)11-10/h3-5,12H,6H2,1-2H3
Formula Weight 180.0609
Chemical Name or Material 2,2-Dimethyl-2,3-dihydro-1-benzofuran-5-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.