missing translation for 'onlineSavingsMsg'
Learn More

2,2’-Dipyridyl, TRC

Product Code. 30138035
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138035 100 g 100g
30010011 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138035 Supplier Toronto Research Chemicals Supplier No. TRCD492735100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-18-7
Molecular Formula C10 H8 N2
Molecular Weight (g/mol) 156.18
Synonym 2,2'-Bipyridyl, 2,2'-Dipyridine, 2,2'-Dipyridyl, 2-(2-Pyridyl)pyridine, Dri-Rx 19LC-E, NSC 1550, NSC 615009, α,α'-Bipyridine, α,α'-Bipyridyl, α,α'-Dipyridine, α,α'-Dipyridyl
IUPAC Name 2-pyridin-2-ylpyridine
SMILES c1ccc(nc1)c2ccccn2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H8N2/c1-3-7-11-9(5-1)10-6-2-4-8-12-10/h1-8H
Formula Weight 156.0687
Chemical Name or Material 2,2'-Bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.