missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Oxybis(N,N-dimethylethanamine) (Bis[2-(dimethylamino)ethyl] Ether; N,N,N',N’-Tetramethyl(oxydiethylene)diamine), Mikromol™

Product Code. 18032490
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18032490

Brand: Mikromol MM0273.170100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,2'-Oxybis(N,N-dimethylethanamine) (Bis[2-(dimethylamino)ethyl] Ether; N,N,N',N’-Tetramethyl(oxydiethylene)diamine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2'-Oxybis(N,N-dimethylethanamine)
CAS 3033-62-3
Analyte or Component Names 2,2'-Oxybis(N,N-dimethylethanamine)
Shipping Condition Room Temperature
Molecular Formula C8 H20 N2 O
Certifications/Compliance ISO 9001
Synonym Ethanamine, 2,2'-oxybis[N,N-dimethyl-, Ethylamine, 2,2'-oxybis[N,N-dimethyl- (6CI,7CI,8CI), 2,2'-Oxybis[N,N-dimethylethanamine], 1,5-Bis(dimethylamino)-3-oxapentane, 2,2'-Oxybis[N,N-dimethylethylamine], A 1, A 133, A 99, A 99 (amine), Addocat 108, BL 11, BL 19, Bis(dimethylaminoethyl) ether, Bis[2-(dimethylamino)ethyl] ether, DY 1, Dabco BL 11, Dabco BL 19, Dabco BL 19I, ET 33B, Jeffcat Z 20, Jeffcat ZF 20, Jeffcat ZM 20, Kalpur PC, Kaolizer 12, Kaolizer 12P, Minico TMDA, N,N,N',N'-Tetramethyl-2,2'-oxybis[ethylamine], N,N,N',N'-Tetramethyl-3-oxapentane-1,5-diamine, N,N,N',N'-Tetramethylbisaminoethyl ether, NP 99, NSC 109887, Niax A 1, Niax A 99, Niax Catalyst A 1, PC CAT NP 133, PC CAT NP 90, PC CAT NP 99, Tetramethyl bis(aminoethyl) ether, Texacat ZF 20, Texacat ZF 22, Toyocat ET, Toyocat ET 33B, Toyocat ETS, ZF 20, 2,2’-Oxybis(N,N-dimethylethanamine), Potassium Clavulanate Impurity M
InChI Formula InChI=1S/C8H20N2O/c1-9(2)5-7-11-8-6-10(3)4/h5-8H2,1-4H3
SMILES CN(C)CCOCCN(C)C
IUPAC Name 2-[2-(dimethylamino)ethoxy]-N,N-dimethyl-ethanamine
Molecular Weight (g/mol) 160.257
Quantity 100 mg
Formula Weight 160.1576 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.