missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11337201 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11337201

Brand: Thermo Scientific Acros 442210010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-02-6
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002447
InChI Key BRRSNXCXLSVPFC-UHFFFAOYSA-N
Synonym 4-pyrogallolcarboxylic acid, pyrogallolcarboxylic acid, 2,3,4-trihydroxybenzoate, unii-ad1id2jf5o, benzoic acid, trihydroxy, benzoic acid, 2,3,4-trihydroxy, pyrogallol-4-carboxylic acid, 2,3,4-trihydroxybenzene carboxylic acid, ad1id2jf5o, pyrogallolcarboxylate
PubChem CID 11874
IUPAC Name 2,3,4-trihydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(O)=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 196°C to 201°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 170.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 3, 4-Trihydroxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4-Trihydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.