missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Trimethylphenol, 98+%, Thermo Scientific Chemicals

Product Code. p-207537 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10593491

Brand: Thermo Scientific Acros 140190050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 697-82-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002228
InChI Key OGRAOKJKVGDSFR-UHFFFAOYSA-N
Synonym isopseudocumenol, phenol, 2,3,5-trimethyl, 1-hydroxy-2,3,5-trimethylbenzene, unii-1s1061zbq1, phenol, 2,3,5 or 3,4,5-trimethyl, pubchem2249, 2,3,5-trimethyphenol, phenol,3,5-trimethyl, 2,3,5-trimethyl-phenol, 6-hydroxypseudocumene
PubChem CID 12769
ChEBI CHEBI:38570
IUPAC Name 2,3,5-trimethylphenol
SMILES CC1=CC(=C(C(=C1)O)C)C

Specifikationer

CAS Min % 98.0
CAS Max % 100.0
Melting Point 92°C to 95°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula (CH3)3C6H2OH
Quantity 5 g
Formula Weight 136.19
Percent Purity ≥98.0%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material 2, 3, 5-Trimethylphenol, 98+%
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 2,3,5-Trimethylphenol
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Faroangivelser
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.