missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide

Brand:  Alfa Aesar™ J66024.06

Additional Details : CAS : 14227-66-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15410497

  • 107.00€ / 5g


Estimated Shipment
11-05-2021
Add to basket

Description

Description

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water. Slightly soluble in ethanol

Notes
Hygroscopic, store away from water/moisture. Keep the container tightly closed. Store away from oxidizing agent. Store under dry inert gas.
Specifications

Chemical Identifiers

14227-66-8
699.453
NLFHLQWXGDPOME-VRECAULFSA-N
11039776
CC(=O)OCC1C(C(C(C(O1)OC2C(OC(C(C2OC(=O)C)OC(=O)C)Br)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
C26H35BrO17
MFCD00069839
cel br heptaacetate, alpha-d-cellobiosyl bromide heptaacetate, cellobiosyl bromide heptaacetate, acetobromo-d-cellobiose, 2,3,6,2',3',4',6'-hepta-o-acetyl-a-d-cellobiosyl bromide, 2r,3r,4s,5r,6s-2-acetoxymethyl-6-2r,3r,4s,5r,6r-4,5-diacetoxy-2-acetoxymethyl-6-bromotetrahydro-2h-pyran-3-yl oxy tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-2r,3r,4s,5r,6r-4,5-bis acetyloxy-2-acetyloxy methyl-6-bromooxan-3-yl oxy oxan-2-yl methyl acetate, alpha-d-glucopyranosyl bromide, 4-o-2,3,4,6-tetra-o-acetyl-beta-d-glucopyranosyl-, triacetate
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-3-yl]oxyoxan-2-yl]methyl acetate

Specifications

2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide
White
5g
C26H35BrO17
cel br heptaacetate, alpha-d-cellobiosyl bromide heptaacetate, cellobiosyl bromide heptaacetate, acetobromo-d-cellobiose, 2,3,6,2',3',4',6'-hepta-o-acetyl-a-d-cellobiosyl bromide, 2r,3r,4s,5r,6s-2-acetoxymethyl-6-2r,3r,4s,5r,6r-4,5-diacetoxy-2-acetoxymethyl-6-bromotetrahydro-2h-pyran-3-yl oxy tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-2r,3r,4s,5r,6r-4,5-bis acetyloxy-2-acetyloxy methyl-6-bromooxan-3-yl oxy oxan-2-yl methyl acetate, alpha-d-glucopyranosyl bromide, 4-o-2,3,4,6-tetra-o-acetyl-beta-d-glucopyranosyl-, triacetate
NLFHLQWXGDPOME-VRECAULFSA-N
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-3-yl]oxyoxan-2-yl]methyl acetate
11039776
Powder
Hygroscopic
180°C to 190°C
14227-66-8
MFCD00069839
Soluble in water; Slightly soluble in ethanol
CC(=O)OCC1C(C(C(C(O1)OC2C(OC(C(C2OC(=O)C)OC(=O)C)Br)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
699.453
699.45
Videos
SDS Documents