missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 15423207
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423207 25 g 25g
15433207 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423207 Supplier Thermo Scientific Alfa Aesar Supplier No. B20165.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69045-84-7
Molecular Formula C6H2Cl2F3N
Molecular Weight (g/mol) 215.98
MDL Number MFCD00042243
InChI Key ABNQGNFVSFKJGI-UHFFFAOYSA-N
Synonym 2,3-dichloro-5-trifluoromethyl pyridine, unii-5f2c8lx8ie, 2,3-dichloro-5-trifluoro methylpyridine, 2,3,5-dctf, 5f2c8lx8ie, 2,3-dichloro-5-trifluoromethyl-pyridine, pyridine, 2,3-dichloro-5-trifluoromethyl, pubchem9237
PubChem CID 112234
IUPAC Name 2,3-dichloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CN=C(Cl)C(Cl)=C1

Specifications

Density 1.54
Boiling Point 79°C to 81°C (20 mmHg)
Flash Point 79°C (175°F)
Refractive Index 1.475
Quantity 25 g
UN Number UN3082
Formula Weight 215.99
Percent Purity 97%
Chemical Name or Material 2,3-Dichloro-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.