missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenoxyacetic acid, 97%

Product Code. 11470753
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470753 5 g 5g
11480753 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470753 Supplier Thermo Scientific Alfa Aesar Supplier No. A13231.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 2976-74-1 | C8H6Cl2O3 | 221.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2976-74-1
Molecular Formula C8H6Cl2O3
Molecular Weight (g/mol) 221.033
MDL Number MFCD00004299
InChI Key RBJIGQRZLITQJG-UHFFFAOYSA-N
Synonym 2,3-dichlorophenoxyacetic acid, 2-2,3-dichlorophenoxy acetic acid, 2,3-dichlorophenoxy acetic acid, 2,3-dichlorophenoxyaceticacid, acetic acid,2-2,3-dichlorophenoxy, acetic acid, 2,3-dichlorophenoxy, pubchem23860, acmc-20a9i3, 2,3-dichloro phenoxyacetic acid, acetic acid,3-dichlorophenoxy
PubChem CID 18105
IUPAC Name 2-(2,3-dichlorophenoxy)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)OCC(=O)O

Specifications

Melting Point 173°C to 175°C
Quantity 5 g
Beilstein 1963475
Formula Weight 221.04
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorophenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.