missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoroanisole, 97+%, Thermo Scientific™

Catalog Number 11489967
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
134364-69-5
C7H6F2O
144.12
MFCD01631562
RDOGTTNFVLSBKG-UHFFFAOYSA-N
2,3-difluoroanisole, 1,2-difluoro-3-methoxy-benzene, 2,3-fluoroanisole, 2,3-difluoro-1-methoxybenzene, benzene, 1,2-difluoro-3-methoxy, 2,3-difluoroanisol, difluro-methoxybenzen, pubchem1969, 2,3-difluoro anisole, acmc-1bwhl
2769344
1,2-difluoro-3-methoxybenzene
COC1=CC=CC(F)=C1F
This item is not returnable. View return policy
134364-69-5
C7H6F2O
144.12
MFCD01631562
RDOGTTNFVLSBKG-UHFFFAOYSA-N
2,3-difluoroanisole, 1,2-difluoro-3-methoxy-benzene, 2,3-fluoroanisole, 2,3-difluoro-1-methoxybenzene, benzene, 1,2-difluoro-3-methoxy, 2,3-difluoroanisol, difluro-methoxybenzen, pubchem1969, 2,3-difluoro anisole, acmc-1bwhl
2769344
1,2-difluoro-3-methoxybenzene
COC1=CC=CC(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Flash Point 62°C (144°F)
Quantity 1 g
Formula Weight 144.12
Percent Purity ≥97%
Chemical Name or Material 2,3-Difluoroanisole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'