missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenylacetic acid, 97%, Thermo Scientific™

Catalog Number p-4228603
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
145689-41-4
C8H6F2O2
172.13
MFCD00040968
UXSQXUSJGPVOKT-UHFFFAOYSA-N
2,3-difluorophenylacetic acid, 2-2,3-difluorophenyl acetic acid, 2,3-difluorophenyl acetic acid, 2,3-difluorophenylaceticacid, benzeneacetic acid, 2,3-difluoro, 2,3-difluorobenzeneacetic acid, rarechem al bo 0630, pubchem1916, acmc-1crot
520772
2-(2,3-difluorophenyl)acetic acid
C1=CC(=C(C(=C1)F)F)CC(=O)O
This item is not returnable. View return policy
145689-41-4
C8H6F2O2
172.13
MFCD00040968
UXSQXUSJGPVOKT-UHFFFAOYSA-N
2,3-difluorophenylacetic acid, 2-2,3-difluorophenyl acetic acid, 2,3-difluorophenyl acetic acid, 2,3-difluorophenylaceticacid, benzeneacetic acid, 2,3-difluoro, 2,3-difluorobenzeneacetic acid, rarechem al bo 0630, pubchem1916, acmc-1crot
520772
2-(2,3-difluorophenyl)acetic acid
C1=CC(=C(C(=C1)F)F)CC(=O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 117.0°C to 119.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 172.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Difluorophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Difluorophenylacetic acid
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'