missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylquinoxaline, 97%

Product Code. 11400927
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11400927 5 g 5g
11410927 25 g 25g
11420927 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11400927

Brand: Thermo Scientific Alfa Aesar B23313.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2379-55-7 | C10H10N2 | 158.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2379-55-7
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00006728
InChI Key FKHNZQFCDGOQGV-UHFFFAOYSA-N
PubChem CID 16925
IUPAC Name 2,3-dimethylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1C

Specifications

Melting Point 102°C to 107°C
Quantity 5 g
Beilstein 114832
Synonym quinoxaline, 2,3-dimethyl, aurora ka-3449, quinoxaline,3-dimethyl, 2,3-dimethyl-quinoxalin, 2,3-dimethyl-quinoxaline, acmc-1cg5s, quinoxaline,2,3-dimethyl, salor-int l498777-1ea, 5-23-07-00267 beilstein handbook reference, chembl67865
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylquinoxaline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.