missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Trifluoromethyl)phenyl]ethylamine, 98%, Thermo Scientific Chemicals

Catalog Number 15411129
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
52516-30-0
C9H10F3N
189.181
MFCD00040756
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
104223
2-[3-(trifluoromethyl)phenyl]ethanamine
C1=CC(=CC(=C1)C(F)(F)F)CCN
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
52516-30-0
C9H10F3N
189.181
MFCD00040756
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
104223
2-[3-(trifluoromethyl)phenyl]ethanamine
C1=CC(=CC(=C1)C(F)(F)F)CCN

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.18 g/mL
Boiling Point 113°C (22 mmHg)
Refractive Index 1.4645
Quantity 25 g
UN Number UN2735
Beilstein 4398117
Sensitivity Light sensitive
Formula Weight 189.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-[3-(Trifluoromethyl)phenyl]ethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'