missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Collidine, 99%, Thermo Scientific Chemicals

Catalog Number p-93088
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
108-75-8
C8H11N
121.18
MFCD00006338
BWZVCCNYKMEVEX-UHFFFAOYSA-N
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
7953
2,4,6-trimethylpyridine
CC1=CC(C)=NC(C)=C1
108-75-8
C8H11N
121.18
MFCD00006338
BWZVCCNYKMEVEX-UHFFFAOYSA-N
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
7953
2,4,6-trimethylpyridine
CC1=CC(C)=NC(C)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 171.0°C to 172.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H2N)(CH3)3
Refractive Index 1.4995 to 1.5015
Quantity 100 mL
Beilstein 20, 250
Fieser 01,155; 13,302
Merck Index 15, 9890
Specific Gravity 0.91
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: soluble in diluted acids,soluble in alcohol,chloroform,benzene and,toluene,miscible with ether
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4, 6-Collidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Collidine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'